2-Chloro-5-nitrobenzonitrile ≥98.0%

Supplier: TCI America

C2026-25G C2026-5G
TCC2026-25GEA 265.44 CAD
TCC2026-25G TCC2026-5G
2-Chloro-5-nitrobenzonitrile ≥98.0%
2-Chloro-5-nitrobenzonitrile

CAS Number: 16588-02-6
MDL Number: MFCD00007292
Molecular Formula: C7H3ClN2O2
Molecular Weight: 182.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 122
Melting point (°C): 107

Formula: C₇H₃ClN₂O₂
Boiling Pt: 122 °C
Melting Pt: 107 °C
MDL Number: MFCD00007292
CAS Number: 16588-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17f647b76f4e57a57d1d78c9440b87ef77ee1dba&vid=655da907d704f08b00d683f9c77c1fb0c5bbdf71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR