2-Chloro-5-methyl-3-nitropyridine ≥98.0%
Supplier: TCI America
Synonyms:
2-Chloro-3-nitro-5-picoline
C2022-5G
C2022-1G
TCC2022-5GEA
194.97
CAD
TCC2022-5G
TCC2022-1G
2-Chloro-5-methyl-3-nitropyridine ≥98.0%
2-Chloro-5-methyl-3-nitropyridine
CAS Number: 23056-40-8
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 47
Other Product Names / Synonyms: 6-Chloro-5-nitro-3-picoline
Formula:
C₆H₅ClN₂O₂ MW: 172.57 g/mol Melting Pt: 47 °C |
MDL Number:
MFCD02070020 CAS Number: 23056-40-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...