Cyclopropanecarboxamide ≥98.0%

Supplier: TCI America

C2010-250G C2010-25G
TCC2010-250GEA 603.24 CAD
TCC2010-250G TCC2010-25G
Cyclopropanecarboxamide ≥98.0%
Cyclopropanecarboxamide

CAS Number: 6228-73-5
Molecular Formula: C4H7NO
Molecular Weight: 85.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 122

Formula: C₄H₇NO
Melting Pt: 122 °C
MDL Number: MFCD00013729
CAS Number: 6228-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ec703c20e286fdb99d70a42b54afddf5399183a&vid=4ec703c20e286fdb99d70a42b54afddf5399183a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR