(±)-3-Chloromandelic acid ≥97.0%

Supplier: TCI America

Synonyms: (±)-3-Chlorophenylglycolic acid, 3-Chloromandelic acid

C2009-25G C2009-5G
TCC2009-25GEA 235.42 CAD
TCC2009-25G TCC2009-5G
(±)-3-Chloromandelic acid ≥97.0%
(±)-3-Chloromandelic acid

CAS Number: 16273-37-3
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 115


Other Product Names / Synonyms: 2-(3-Chlorophenyl)-2-hydroxyacetic Acid

Formula: C₈H₇ClO₃
MW: 186.59
Melting Pt: 115 °C
MDL Number: MFCD00829409
CAS Number: 16273-37-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e46a9bfbe0201a45c40b42001560607afa86bb59&vid=e46a9bfbe0201a45c40b42001560607afa86bb59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR