2-Chloroveratraldehyde ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-3,4-dimethoxybenzaldehyde

C1997-25G C1997-5G
TCC1997-25GEA 153.26 CAD
TCC1997-25G TCC1997-5G
2-Chloroveratraldehyde ≥98.0%
2-Chloroveratraldehyde

CAS Number: 5417-17-4
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 71


Other Product Names / Synonyms: 2-Chloroveratraldehyde

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Melting Pt: 71 °C
MDL Number: MFCD00084942
CAS Number: 5417-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2181afcf1c852311c03bbaa819b2c9922504d6d6&vid=d258348896bcaa2eb5ee7df4c6328b4ae4fb4eac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR