O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate ≥98.0% (by HPLC)

Supplier: TCI America

C1988-1G C1988-5G
TCC1988-1GEA 33.18 CAD
TCC1988-1G TCC1988-5G
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate ≥98.0% (by HPLC)
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate

CAS Number: 330645-87-9
Molecular Formula: C11H15ClF6N5OP
Molecular Weight: 413.69
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: HCTU

Formula: C₁₁H₁₅ClF₆N₅OP
MDL Number: MFCD12031645
CAS Number: 330645-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27e2926927906831b1a03d342f5cf3cb14cc6eb&vid=dd6408cd09905a62885ffb6e3c3792a30f4ead31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR