5-Chloro-2-hydroxy-3-nitropyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Chloro-3-nitropyridin-2-ol

C1972-5G C1972-25G
TCC1972-5GEA 224.83 CAD
TCC1972-5G TCC1972-25G
5-Chloro-2-hydroxy-3-nitropyridine ≥98.0% (by titrimetric analysis)
5-Chloro-2-hydroxy-3-nitropyridine

CAS Number: 21427-61-2
Molecular Formula: C5H3ClN2O3
Molecular Weight: 174.54
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 236


Other Product Names / Synonyms: 5-Chloro-3-nitro-2-pyridinol

Formula: C₅H₃ClN₂O₃
MW: 174.54
Melting Pt: 236 °C
MDL Number: MFCD00114884
CAS Number: 21427-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=211462b000e35bf2b50c00f73a80007a3e991d3f&vid=211462b000e35bf2b50c00f73a80007a3e991d3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR