6-Chlorochroman-4-one ≥98.0%
Supplier: TCI America
C1968-5G
C1968-25G
TCC1968-5GEA
133.83
CAD
TCC1968-5G
TCC1968-25G
6-Chlorochroman-4-one ≥98.0%
6-Chlorochroman-4-one
CAS Number: 37674-72-9
Molecular Formula: C9H7ClO2
Molecular Weight: 182.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 101
Formula:
C₉H₇ClO₂ Melting Pt: 101 °C |
MDL Number:
MFCD00024070 CAS Number: 37674-72-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...