N-(4-Chlorophenyl)phthalimide ≥98.0%
Supplier: TCI America
Synonyms:
2-(4-Chlorophenyl)-1H-isoindole-1,3(2H)-dione
C1958-5G
C1958-25G
TCC1958-5GEA
151.68
CAD
TCC1958-5G
TCC1958-25G
N-(4-Chlorophenyl)phthalimide ≥98.0%
N-(4-Chlorophenyl)phthalimide
CAS Number: 7386-21-2
Molecular Formula: C14H8ClNO2
Molecular Weight: 257.67
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 198
Formula:
C₁₄H₈ClNO₂ MW: 257.68 Melting Pt: 198 °C |
MDL Number:
MFCD00023048 CAS Number: 7386-21-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...