(6-Chloro-3-pyridinyl)methanol ≥98.0%

Supplier: TCI America

Synonyms: 6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine

C1897-25G C1897-5G
TCC1897-25GEA 317.42 CAD
TCC1897-25G TCC1897-5G
(6-Chloro-3-pyridinyl)methanol ≥98.0%
(6-Chloro-3-pyridinyl)methanol

CAS Number: 21543-49-7
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Chloro-5-(hydroxymethyl)pyridine

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 54 °C
Flash Pt: 110 °C
MDL Number: MFCD01075226
CAS Number: 21543-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=397c0fc79fd296bd5061535df9331bf8969a3191&vid=397c0fc79fd296bd5061535df9331bf8969a3191&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR