2-Chloro-3-picoline ≥98.0%
Supplier: TCI America
C1885-25G
C1885-5G
TCC1885-25GEA
126.4
CAD
TCC1885-25G
TCC1885-5G
2-Chloro-3-picoline ≥98.0%
2-Chloro-3-picoline
CAS Number: 18368-76-8
Molecular Formula: C6H6ClN
Molecular Weight: 127.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 79
Specific Gravity (20/20): 1.17
Other Product Names / Synonyms: 2-Chloro-3-picoline
Formula:
C₆H₆ClN Boiling Pt: 193 °C Density: 1.17 (20 °C) Flash Pt: 79 °C |
MDL Number:
MFCD00234177 CAS Number: 18368-76-8 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...