5-Chloroindole-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1868-25G C1868-5G
TCC1868-25GEA 538.78 CAD
TCC1868-25G TCC1868-5G
5-Chloroindole-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Chloroindole-2-carboxylic acid

CAS Number: 10517-21-2
MDL Number: MFCD00005613
Molecular Formula: C9H6ClNO2
Molecular Weight: 195.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 290

Formula: C₉H₆ClNO₂
Melting Pt: 290 °C
MDL Number: MFCD00005613
CAS Number: 10517-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f3bf3e42fb770f33fcf93c9b6c6640d89d8898c&vid=4f3bf3e42fb770f33fcf93c9b6c6640d89d8898c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR