5-Bromo-2-chlorobenzotrifluoride ≥98.0%

Supplier: TCI America

Synonyms: 1-Bromo-4-chloro-3-(trifluoromethyl)benzene, 5-Bromo-2-chlorobenzotrifluoride, 4-Bromo-1-chloro-2-(trifluoromethyl)benzene, 5-Bromo-2-chloro-α,α,α-trifluorotoluene

C1846-25G C1846-5G
TCC1846-25GEA 137.46 CAD
TCC1846-25G TCC1846-5G
5-Bromo-2-chlorobenzotrifluoride ≥98.0%
5-Bromo-2-chlorobenzotrifluoride

CAS Number: 445-01-2
Molecular Formula: C7H3BrClF3
Molecular Weight: 259.45
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Melting point (°C): -21
Flash Point (°C): 81
Specific Gravity (20/20): 1.75


Other Product Names / Synonyms: 4-Bromo-1-chloro-2-(trifluoromethyl)benzene, 5-Bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₃BrClF₃
Boiling Pt: 195 °C
Melting Pt: –21 °C
Density: 1.75 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00000601
CAS Number: 445-01-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR