Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥93.0%
Supplier: TCI America
Synonyms:
[Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer
CAS Number: 12112-67-3
MDL Number: MFCD00012414
Molecular Formula: C16H24Cl2Ir2
Molecular Weight: 671.70
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Melting point (°C): 205
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction, Metal Catalysts ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Bis(1,5-cyclooctadiene)diiridium(I) Dichloride
Formula:
C₁₆H₂₄Cl₂Ir₂ MW: 671.70 g/mol Melting Pt: 205 °C |
MDL Number:
MFCD00012414 CAS Number: 12112-67-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...