3-Chloro-4-methoxybenzaldehyde ≥98.0%
Supplier: TCI America
C1797-1G
C1797-5G
TCC1797-1GEA
80.58
CAD
TCC1797-1G
TCC1797-5G
3-Chloro-4-methoxybenzaldehyde ≥98.0%
3-Chloro-4-methoxybenzaldehyde
CAS Number: 4903-09-7
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 58
Other Product Names / Synonyms: 3-Chloro-4-methoxybenzaldehyde
Formula:
C₈H₇ClO₂ Melting Pt: 58 °C |
MDL Number:
MFCD00016601 CAS Number: 4903-09-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...