5-Chloro-2-nitrodiphenylamine ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloro-2-nitro-N-phenylaniline

C1785-5G C1785-25G
TCC1785-5GEA 65.89 CAD
TCC1785-5G TCC1785-25G
5-Chloro-2-nitrodiphenylamine ≥98.0%
5-Chloro-2-nitrodiphenylamine

CAS Number: 25781-92-4
MDL Number: MFCD00007287
Molecular Formula: C12H9ClN2O2
Molecular Weight: 248.67
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 114

Formula: C₁₂H₉ClN₂O₂
MW: 248.6700 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00007287
CAS Number: 25781-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03b4e61c0fee6df8ab49af81825fd32bef4f4ce8&vid=03b4e61c0fee6df8ab49af81825fd32bef4f4ce8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR