2-Chloro-4,6-difluoroaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1777-5G C1777-1G
TCC1777-5GEA 80.58 CAD
TCC1777-5G TCC1777-1G
2-Chloro-4,6-difluoroaniline ≥98.0% (by GC, titration analysis)
2-Chloro-4,6-difluoroaniline

CAS Number: 36556-56-6
MDL Number: MFCD00142892
Molecular Formula: C6H4ClF2N
Molecular Weight: 163.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4,6-Difluoro-2-chloroaniline

Formula: C₆H₄ClF₂N
MW: 163.55 g/mol
MDL Number: MFCD00142892
CAS Number: 36556-56-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b9277fa3bd729432c0c87532cd3fa822e44156a&vid=9b9277fa3bd729432c0c87532cd3fa822e44156a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR