5-Chloro-2-methylindole ≥98.0%
Supplier: TCI America
C1761-5G
C1761-1G
TCC1761-5GEA
312.84
CAD
TCC1761-5G
TCC1761-1G
5-Chloro-2-methylindole ≥98.0%
5-Chloro-2-methylindole
CAS Number: 1075-35-0
MDL Number: MFCD00005619
Molecular Formula: C9H8ClN
Molecular Weight: 165.62
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 117
Other Product Names / Synonyms: 2-Methyl-5-chloroindole
Formula:
C₉H₈ClN MW: 165.62 Boiling Pt: 140 °C Melting Pt: 129 |
MDL Number:
MFCD00005619 CAS Number: 1075-35-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...