3-Chloro-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C1760-5G
C1760-1G
TCC1760-5GEA
130.51
CAD
TCC1760-5G
TCC1760-1G
3-Chloro-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-4-fluorophenylboronic acid
CAS Number: 144432-85-9
MDL Number: MFCD00051800
Molecular Formula: C6H5BClFO2
Molecular Weight: 174.36
Form: Crystal
Color: White
Melting point (°C): 247
Other Product Names / Synonyms: 3-Chloro-4-fluorobenzeneboronic Acid
Formula:
C₆H₅BClFO₂ Melting Pt: 247 °C |
MDL Number:
MFCD00051800 CAS Number: 144432-85-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...