2-Chloro-5-ethylpyrimidine ≥98.0%

Supplier: TCI America

C1754-5G C1754-1G
TCC1754-5GEA 112.18 CAD
TCC1754-5G TCC1754-1G
2-Chloro-5-ethylpyrimidine ≥98.0%
2-Chloro-5-ethylpyrimidine

CAS Number: 111196-81-7
MDL Number: MFCD00799503
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 100
Specific Gravity (20/20): 1.19

Formula: C₆H₇ClN₂
Boiling Pt: 86 °C
Density: 1.19 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00799503
CAS Number: 111196-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=749a0a8b3e0d02aea8599f0c24227fe5f7c4fcd2&vid=749a0a8b3e0d02aea8599f0c24227fe5f7c4fcd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR