Chloromethyl polystyrene resin 0,8 - 1,2 mmol/g 0.8-1.2 mmol/g, cross-linked with 2% DVB (100-200mesh)

Supplier: TCI America

Synonyms: Chloromethylated styrene-divinylbenzene copolymer, Merrifield Resin

C1751-5G C1751-25G
TCC1751-5GEA 51.44 CAD
TCC1751-5G TCC1751-25G
Chloromethyl polystyrene resin 0,8 - 1,2 mmol/g 0.8-1.2 mmol/g, cross-linked with 2% DVB (100-200mesh)
Chloromethyl polystyrene resin

MDL Number: MFCD00146406


Other Product Names / Synonyms: Merrifield Resin

MDL Number: MFCD00146406
CAS Number: 55844-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb32bf0814fa147b4d02cc7cbab0afffc1946339&vid=eb32bf0814fa147b4d02cc7cbab0afffc1946339&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR