Chloromethyl polystyrene resin 2,0-3,0 mmol/g 2.0-3.0 mmol/g, cross-linked with 1% DVB (100-200mesh)

Supplier: TCI America

Synonyms: Chloromethylated styrene-divinylbenzene copolymer, Merrifield Resin

C1750-5G C1750-25G
TCC1750-5GEA 102.84 CAD
TCC1750-5G TCC1750-25G
Chloromethyl polystyrene resin 2,0-3,0 mmol/g 2.0-3.0 mmol/g, cross-linked with 1% DVB (100-200mesh)
Chloromethyl polystyrene resin

MDL Number: MFCD00146406
Form: Crystal
Color: White


Other Product Names / Synonyms: Merrifield Resin

MDL Number: MFCD00146406
CAS Number: 55844-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d57621844aa236c2a9bdae078b50745f726c8f81&vid=d57621844aa236c2a9bdae078b50745f726c8f81&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR