Ethyl-3-chlorobenzoate ≥94.0%

Supplier: TCI America

C1737-25G C1737-5G
TCC1737-25GEA 102.54 CAD
TCC1737-25G TCC1737-5G
Ethyl-3-chlorobenzoate ≥94.0%
Ethyl 3-chlorobenzoate

CAS Number: 1128-76-3
MDL Number: MFCD00013634
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: 3-Chlorobenzoic Acid Ethyl Ester

Formula: C₉H₉ClO₂
Density: 1.19 (20 °C)
MDL Number: MFCD00013634
CAS Number: 1128-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e53f3f5590e262261b7e8fab79a88dc0465c21c1&vid=e53f3f5590e262261b7e8fab79a88dc0465c21c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR