Methyl coumalate ≥96.0%
Supplier: TCI America
C1734-25G
C1734-5G
TCC1734-25GEA
413.17
CAD
TCC1734-25G
TCC1734-5G
Methyl coumalate ≥96.0%
Methyl coumalate
CAS Number: 6018-41-3
MDL Number: MFCD00010120
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 68
Other Product Names / Synonyms: Coumalic Acid Methyl Ester, 2-Oxopyran-5-carboxylic Acid Methyl Ester, Methyl 2-Oxopyran-5-carboxylate, 2-Pyrone-5-carboxylic Acid Methyl Ester, Methyl 2-Pyrone-5-carboxylate
Formula:
C₇H₆O₄ Boiling Pt: 260 °C Melting Pt: 68 °C |
MDL Number:
MFCD00010120 CAS Number: 6018-41-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...