Methyl-2-chloro-5-nitrobenzoate ≥98.0%
Supplier: TCI America
C1732-25G
C1732-5G
TCC1732-25GEA
148.52
CAD
TCC1732-25G
TCC1732-5G
Methyl-2-chloro-5-nitrobenzoate ≥98.0%
Methyl 2-chloro-5-nitrobenzoate
CAS Number: 6307-82-0
MDL Number: MFCD00184587
Molecular Formula: C8H6ClNO4
Molecular Weight: 215.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70
Other Product Names / Synonyms: 2-Chloro-5-nitrobenzoic Acid Methyl Ester
Formula:
C₈H₆ClNO₄ Melting Pt: 70 °C |
MDL Number:
MFCD00184587 CAS Number: 6307-82-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...