N-Carbobenzyloxy-D-proline ≥98.0%

Supplier: TCI America

Synonyms: (R)-1-((Benzyloxy)carbonyl)pyrrolidine-2-carboxylic acid, Z-D-Pro-OH, N-Cbz-D-proline, Cbz-D-Pro-OH, ((Benzyloxy)carbonyl)-D-proline, N-Benzyloxycarbonyl-D-proline

C1730-5G C1730-25G
TCC1730-5GEA 69.36 CAD
TCC1730-5G TCC1730-25G
N-Carbobenzyloxy-D-proline ≥98.0%
N-Carbobenzyloxy-D-proline

CAS Number: 6404-31-5
MDL Number: MFCD00063228
Molecular Formula: C13H15NO4
Molecular Weight: 249.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 76
Specific rotation [a]20/D: 40 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-proline, Z-D-Pro-OH

Formula: C₁₃H₁₅NO₄
MW: 249.3 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00063228
CAS Number: 6404-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54ce1e5f6081f3589736590f16d5197905816206&vid=54ce1e5f6081f3589736590f16d5197905816206&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR