N-Carbobenzoxy-L-tyrosine ≥98.0%

Supplier: TCI America

Synonyms: Z-Tyr-OH, N-Cbz-L-tyrosine, Cbz-Tyr-OH

C1729-25G C1729-5G
TCC1729-25GEA 235.42 CAD
TCC1729-25G TCC1729-5G
N-Carbobenzoxy-L-tyrosine ≥98.0%
N-Carbobenzoxy-L-tyrosine

CAS Number: 1164-16-5
MDL Number: MFCD00037180
Molecular Formula: C17H17NO5
Molecular Weight: 315.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 95
Specific rotation [a]20/D: 10 deg (C=1, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-L-tyrosine, Z-Tyr-OH

Formula: C₁₇H₁₇NO₅
MW: 315.33 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00037180
CAS Number: 1164-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e10760397e7284c6920cf0f02da24e5be66c7a4&vid=3e10760397e7284c6920cf0f02da24e5be66c7a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR