5-Chlorothiophene-2-carboxaldehyde ≥95.0%

Supplier: TCI America

C1722-5G C1722-1G
TCC1722-5GEA 173.48 CAD
TCC1722-5G TCC1722-1G
5-Chlorothiophene-2-carboxaldehyde ≥95.0%
5-Chlorothiophene-2-carboxaldehyde

CAS Number: 7283-96-7
MDL Number: MFCD00047090
Molecular Formula: C5H3ClOS
Molecular Weight: 146.59
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 97
Specific Gravity (20/20): 1.39


Other Product Names / Synonyms: 2-Chloro-5-formylthiophene

Formula: C₅H₃ClOS
Density: 1.39 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00047090
CAS Number: 7283-96-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fde912cf982fe66c03b886871d0f08ba52b85bb7&vid=fde912cf982fe66c03b886871d0f08ba52b85bb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR