Ethyl-(S)-4-Chloro-3-hydroxybutyrate ≥95.0%

Supplier: TCI America

C1717-25G C1717-5G
TCC1717-25GEA 135.88 CAD
TCC1717-25G TCC1717-5G
Ethyl-(S)-4-Chloro-3-hydroxybutyrate ≥95.0%
Ethyl-(S)-4-Chloro-3-hydroxybutyrate

CAS Number: 86728-85-0
MDL Number: MFCD00211241
Molecular Formula: C6H11ClO3
Molecular Weight: 166.60
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 110
Specific Gravity (20/20): 1.19
Specific rotation [a]20/D: -14 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester

Formula: C₆H₁₁ClO₃
Boiling Pt: 96 °C
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00211241
CAS Number: 86728-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0793048e6031f23e829190456c8048ead144460f&vid=0793048e6031f23e829190456c8048ead144460f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR