4-Chloro-3-nitrobenzyl alcohol ≥98.0%
Supplier: TCI America
C1712-25G
C1712-5G
TCC1712-25GEA
327.06
CAD
TCC1712-25G
TCC1712-5G
4-Chloro-3-nitrobenzyl alcohol ≥98.0%
4-Chloro-3-nitrobenzyl alcohol
CAS Number: 55912-20-4
MDL Number: MFCD00007086
Molecular Formula: C7H6ClNO3
Molecular Weight: 187.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 66
Formula:
C₇H₆ClNO₃ Melting Pt: 66 °C |
MDL Number:
MFCD00007086 CAS Number: 55912-20-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...