2-Chlorophenylboronic acid ≥98% (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid

C1705-5G C1705-1G
TCC1705-5GEA 53.72 CAD
TCC1705-5G TCC1705-1G
2-Chlorophenylboronic acid ≥98% (contains varying amounts of Anhydride)
2-Chlorophenylboronic acid

CAS Number: 3900-89-8
MDL Number: MFCD00674012
Molecular Formula: C6H6BClO2
Molecular Weight: 156.37
Form: Crystal
Color: White


Other Product Names / Synonyms: 2-Chlorobenzeneboronic Acid

Formula: C₆H₆BClO₂
MW: 156.38
MDL Number: MFCD00674012
CAS Number: 3900-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac80265b7b0da33ecb7286140b4d14b9affb6fa6&vid=ac80265b7b0da33ecb7286140b4d14b9affb6fa6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR