Chloromethyl polystyrene resin 1,6 - 1,8 mmol/g 1.6-1.8 mmol/g, cross-linked with 1% DVB (200-400mesh)

Supplier: TCI America

Synonyms: Chloromethylated styrene-divinylbenzene copolymer, Merrifield Resin

C1643-25G C1643-5G
TCC1643-025GEA 342.86 CAD
TCC1643-025G TCC1643-005G
Chloromethyl polystyrene resin 1,6 - 1,8 mmol/g 1.6-1.8 mmol/g, cross-linked with 1% DVB (200-400mesh)
Chloromethyl polystyrene resin

MDL Number: MFCD00146406


Other Product Names / Synonyms: Merrifield Resin

MDL Number: MFCD00146406
CAS Number: 55844-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ecad6e709206fe787d4af4ca4a2a356beb4abfc&vid=6ecad6e709206fe787d4af4ca4a2a356beb4abfc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR