Chloromethyl polystyrene resin 0.8 - 1.3 mmol/g, cross-linked with 1% DVB (200-400mesh)

Supplier: TCI America

Synonyms: Chloromethylated styrene-divinylbenzene copolymer, Merrifield Resin

C1642-5G C1642-25G
TCC1642-005GEA 143.78 CAD
TCC1642-005G TCC1642-025G
Chloromethyl polystyrene resin 0.8 - 1.3 mmol/g, cross-linked with 1% DVB (200-400mesh)
Chloromethyl polystyrene resin

MDL Number: MFCD00146406


Other Product Names / Synonyms: Merrifield Resin

MDL Number: MFCD00146406
CAS Number: 55844-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbb1cd3dc4f0c0161c91bcd7af4cceea6b0dbab0&vid=dbb1cd3dc4f0c0161c91bcd7af4cceea6b0dbab0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR