N-Cbz-D-phenylalaninol ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Benzyloxycarbonyl-D-phenylalaninol, Z-D-Phenylalaninol, Cbz-D-Phenylalaninol

C1609-1G C1609-5G
TCC1609-001GEA 64.46 CAD
TCC1609-001G TCC1609-005G
N-Cbz-D-phenylalaninol ≥97.0% (by HPLC)
N-Cbz-D-phenylalaninol

CAS Number: 58917-85-4
MDL Number: MFCD00191193
Molecular Formula: C17H19NO3
Molecular Weight: 285.34
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 93
Specific rotation [a]20/D: 42 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-N-Carbobenzoxy-2-amino-3-phenyl-1-propanol, (R)-N-Cbz-2-amino-3-phenyl-1-propanol, N-Cbz-D-phenylalaninol

Formula: C₁₇H₁₉NO₃
MW: 285.3 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00191193
CAS Number: 58917-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d653d2861281a9d0a6e4625e2460d3197d60dd6d&vid=d653d2861281a9d0a6e4625e2460d3197d60dd6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR