Dibenzyl carbonate ≥98.0%
Supplier: TCI America
C1600-5G
C1600-1G
TCC1600-5GEA
228.31
CAD
TCC1600-5G
TCC1600-001G
Dibenzyl carbonate ≥98.0%
Dibenzyl carbonate
CAS Number: 3459-92-5
MDL Number: MFCD00014436
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 30
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Carbonic Acid Dibenzyl Ester
Formula:
C₁₅H₁₄O₃ Flash Pt: 113 °C |
MDL Number:
MFCD00014436 CAS Number: 3459-92-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...