Sodium-4-chlorobenzenesulfinate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1559-25G C1559-5G
TCC1559-25GEA 169.06 CAD
TCC1559-25G TCC1559-005G
Sodium-4-chlorobenzenesulfinate ≥98.0% (by titrimetric analysis)
Sodium-4-chlorobenzenesulfinate

CAS Number: 14752-66-0
MDL Number: MFCD00035602
Molecular Formula: C6H5ClO2S
Molecular Weight: 198.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4-Chlorobenzenesulfinic Acid Sodium Salt

Formula: C₆H₅ClO₂S
MDL Number: MFCD00035602
CAS Number: 14752-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0493218d5c41820b3d5c0e15a160244abcaae2c1&vid=f46f9bc4f25d4be4c9cf4a341db12e24df17588a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR