N-Carbobenzoxyhydroxylamine ≥97.0%

Supplier: TCI America

Synonyms: N-Hydroxycarbamic acid Benzyl Ester, N-Cbz-hydroxylamine, Benzyl N-Hydroxycarbamate, Benzylhydroxycarbamate

C1557-5G C1557-25G
TCC1557-005GEA 117.39 CAD
TCC1557-005G TCC1557-25G
N-Carbobenzoxyhydroxylamine ≥97.0%
N-Carbobenzoxyhydroxylamine

CAS Number: 3426-71-9
MDL Number: MFCD00010642
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 70
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Benzyl N-Hydroxycarbamate, N-Cbz-hydroxylamine, N-Hydroxycarbamic Acid Benzyl Ester

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00010642
CAS Number: 3426-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=751065cb80459777400d4ade8b9b77d986367e1f&vid=751065cb80459777400d4ade8b9b77d986367e1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR