4-Cyano-4'-pentyloxybiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4-Amyloxy-4'-cyanobiphenyl, 4-Cyano-4'-n-pentyloxybiphenyl, 5OCB, 4'-Amyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-[1,1'-biphenyl]-4-carbonitrile

C1551-1G C1551-5G
TCC1551-001GEA 72.68 CAD
TCC1551-001G TCC1551-005G
4-Cyano-4'-pentyloxybiphenyl ≥98.0%
4-Cyano-4'-pentyloxybiphenyl

CAS Number: 52364-71-3
MDL Number: MFCD00074879
Molecular Formula: C18H19NO
Molecular Weight: 265.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 52
Flash Point (°C): 110


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4'-Amyloxy-4-biphenylcarbonitrile, 4-Amyloxy-4'-cyanobiphenyl, 4'-Pentyloxy-4-biphenylcarbonitrile, 5OCB

Formula: C₁₈H₁₉NO
MW: 265.35 g/mol
Melting Pt: 52 °C
Flash Pt: 110 °C
MDL Number: MFCD00074879
CAS Number: 52364-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0e9a703e69c1817a4f4f5fc4d2daf08e4c7f66d&vid=c0e9a703e69c1817a4f4f5fc4d2daf08e4c7f66d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR