4-Cyano-4'-pentylbiphenyl ≥98.0%

Supplier: TCI America

C1550-5G C1550-25G C1550-1G
TCC1550-005GEA 305.73 CAD
TCC1550-005G TCC1550-25G TCC1550-001G
4-Cyano-4'-pentylbiphenyl ≥98.0%
4-Cyano-4'-pentylbiphenyl

CAS Number: 40817-08-1
MDL Number: MFCD00036350
Molecular Formula: C18H19N
Molecular Weight: 249.36
Purity/Analysis Method: >98.0% (GC)
Form: Liquid
Boiling point (°C): 215
Flash Point (°C): 110
Specific Gravity (20/20): 1.03


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4'-Amyl-4-biphenylcarbonitrile, 4-Amyl-4'-cyanobiphenyl, 4'-Pentyl-4-biphenylcarbonitrile, 5CB

Formula: C₁₈H₁₉N
Boiling Pt: 215 °C
Density: 1.03 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00036350
CAS Number: 40817-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9522baabcf7a03b073c1e49855a3a8a9029b2e50&vid=9522baabcf7a03b073c1e49855a3a8a9029b2e50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR