N-(2-Cyanoethyl)glycine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(2-Cyanoethyl)aminoacetic Acid

C1525-5G C1525-25G
TCC1525-5GEA 50.56 CAD
TCC1525-5G TCC1525-025G
N-(2-Cyanoethyl)glycine ≥98.0% (by titrimetric analysis)
N-(2-Cyanoethyl)glycine

CAS Number: 3088-42-4
MDL Number: MFCD00066903
Molecular Formula: C5H8N2O2
Molecular Weight: 128.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-(2-Cyanoethyl)aminoacetic Acid

Formula: C₅H₈N₂O₂
MW: 128.13 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00066903
CAS Number: 3088-42-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b09ff0ac4161380124742b90bcc7084431665500&vid=b09ff0ac4161380124742b90bcc7084431665500&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR