Croconic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,5-Dihydroxy-4-cyclopentene-1,2,3-trione, 4,5-Dihydroxycyclopent-4-ene-1,2,3-trione

C1483-1G C1483-5G
TCC1483-1GEA 297.04 CAD
TCC1483-1G TCC1483-5G
Croconic acid ≥98.0% (by HPLC)
Croconic acid

CAS Number: 488-86-8
MDL Number: MFCD00181389
Molecular Formula: C5H2O5
Molecular Weight: 142.07
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 114


Other Product Names / Synonyms: 4,5-Dihydroxy-4-cyclopentene-1,2,3-trione

Formula: C₅H₂O₅
MW: 142.07 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00181389
CAS Number: 488-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e48a4d437dc8e04e9832dc979716ca277641393c&vid=e48a4d437dc8e04e9832dc979716ca277641393c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR