Ethyl (R)-(-)-4-cyano-3-hydroxybutyate ≥97.0%

Supplier: TCI America

Synonyms: (R)-4-Cyano-3-hydroxybutyric acid ethyl ester, BUTANOIC ACID, 4-CYANO-3-HYDROXY-, ETHYL ESTER, (3R)-, Ethyl (R)-4-cyano-3-hydroxybutanoate, (R)-ethyl 4-cyano-3-hydroxybutanoate

C1474-1G C1474-25G C1474-5G
TCC1474-1GEA 26.86 CAD
TCC1474-1G TCC1474-25G TCC1474-5G
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate ≥97.0%
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate

CAS Number: 141942-85-0
MDL Number: MFCD00270839
Molecular Formula: C7H11NO3
Molecular Weight: 157.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 270
Flash Point (°C): 110
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-4-Cyano-3-hydroxybutyric Acid Ethyl Ester

Formula: C₇H₁₁NO₃
MW: 157.169 g/mol
Boiling Pt: 270 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00270839
CAS Number: 141942-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcbefbc9da68d4b854bd2da045b3ad445d137ad0&vid=dcbefbc9da68d4b854bd2da045b3ad445d137ad0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR