2-Coumaranone ≥98.0%

Supplier: TCI America

C1445-25G C1445-5G
TCC1445-025GEA 293.88 CAD
TCC1445-025G TCC1445-5G
2-Coumaranone ≥98.0%
2-Coumaranone

CAS Number: 553-86-6
MDL Number: MFCD00005856
Molecular Formula: C8H6O2
Molecular Weight: 134.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 244
Melting point (°C): 50
Flash Point (°C): 110


Other Product Names / Synonyms: 2(3H)-Benzofuranone, Isocoumaranone

Formula: C₈H₆O₂
Boiling Pt: 244 °C
Melting Pt: 50 °C
Flash Pt: 110 °C
MDL Number: MFCD00005856
CAS Number: 553-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19db0a76e0cf688bcb0f3126097a8396717bece8&vid=19db0a76e0cf688bcb0f3126097a8396717bece8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR