Cyclohexyldiphenylphosphine ≥97.0%

Supplier: TCI America

Synonyms: Diphenylcyclohexylphosphine

C1429-25G C1429-5G
TCC1429-25GEA 366.56 CAD
TCC1429-25G TCC1429-005G
Cyclohexyldiphenylphosphine ≥97.0%
Cyclohexyldiphenylphosphine

CAS Number: 6372-42-5
MDL Number: MFCD00046360
Molecular Formula: C18H21P
Molecular Weight: 268.34
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 60


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Diphenylphosphinocyclohexane

Formula: C₁₈H₂₁P
MW: 268.33 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00046360
CAS Number: 6372-42-5

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR