Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
C1343-500G
C1343-25G
TCC1343-500GEA
407.64
CAD
TCC1343-500G
TCC1343-025G
Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate ≥98.0%
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate
CAS Number: 6289-46-9
MDL Number: MFCD00001607
Molecular Formula: C10H12O6
Molecular Weight: 228.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 156
Formula:
C₁₀H₁₂O₆ MW: 228.2 g/mol Melting Pt: 156 °C |
MDL Number:
MFCD00001607 CAS Number: 6289-46-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...