(1S,2R)-(-)-10,2-Camphorsultam ≥98.0%

Supplier: TCI America

Synonyms: (-)-10,2-Camphorsultam, (2R)-(-)-Bornane-10,2-sultam, (1S,5R)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.0(1,5)]decane 3,3-dioxide, (1S)-(−)-2,10-Camphorsultam, [3aS-(3aalpha,6alpha,7abeta)]-Hexahydro-8,8-dimethyl-2,2-dioxide-3H-3a,6-methano-2,1-benzisothiazole, (3aS,6R,7aR)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (3aS,6S,7aS)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (1S,2R,4R)-2,10-camphorsultam, (-)-exo-10,2-Bornanesultam, (2R)-Bornane-10,2-sultam

C1325-1G C1325-5G
TCC1325-001GEA 41.08 CAD
TCC1325-001G TCC1325-5G
(1S,2R)-(-)-10,2-Camphorsultam ≥98.0%
(1S,2R)-(-)-10,2-Camphorsultam

CAS Number: 94594-90-8
MDL Number: MFCD00066271
Molecular Formula: C10H17NO2S
Molecular Weight: 215.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 182
Specific rotation [a]20/D: -31 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Chiral Auxiliaries, Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: (2R)-Bornane-10,2-sultam, (1S,5R)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.0(1,5)]decane 3,3-Dioxide

Formula: C₁₀H₁₇NO₂S
MW: 215.32 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00066271
CAS Number: 94594-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de23caa4536181aab19b42ee795afea381829e52&vid=de23caa4536181aab19b42ee795afea381829e52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR