1-Cyclohexene-1-carboxylic acid ≥97.0%

Supplier: TCI America

Synonyms: Cyclohex-1-enecarboxylic acid

C1296-5G C1296-1G
TCC1296-5GEA 227.52 CAD
TCC1296-5G TCC1296-001G
1-Cyclohexene-1-carboxylic acid ≥97.0%
1-Cyclohexene-1-carboxylic acid

CAS Number: 636-82-8
MDL Number: MFCD00001545
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 135
Freezing point (°C): 37


Other Product Names / Synonyms: 3,4,5,6-Tetrahydrobenzoic Acid

Formula: C₇H₁₀O₂
Boiling Pt: 135 °C
MDL Number: MFCD00001545
CAS Number: 636-82-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f664596eb7de7f59eb065c1ef3e5e7739d3c5cef&vid=f664596eb7de7f59eb065c1ef3e5e7739d3c5cef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR