2-(1-Cyclohexenyl)cyclohexanone ≥85.0%

Supplier: TCI America

C1160-25ML C1160-500ML
TCC1160-025MLEA 75.84 CAD
TCC1160-025ML TCC1160-500ML
2-(1-Cyclohexenyl)cyclohexanone ≥85.0%
2-(1-Cyclohexenyl)cyclohexanone

CAS Number: 1502-22-3
MDL Number: MFCD00013796
Molecular Formula: C12H18O
Molecular Weight: 178.28
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 275
Flash Point (°C): 134
Specific Gravity (20/20): 1.00

Formula: C₁₂H₁₈O
Boiling Pt: 275 °C
Density: 1.00 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00013796
CAS Number: 1502-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5679bebf70312c89919568af7374bce261a051aa&vid=5679bebf70312c89919568af7374bce261a051aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR