Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers) ≥92.0%

Supplier: TCI America

Synonyms: Dimethyl cyclohexane-1,4-dicarboxylate

C1154-25G C1154-500G
TCC1154-025GEA 47.4 CAD
TCC1154-025G TCC1154-500G
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers) ≥92.0%
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers)

CAS Number: 94-60-0
MDL Number: MFCD00001460
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Boiling point (°C): 263
Flash Point (°C): 94
Freezing point (°C): 13


Other Product Names / Synonyms: 1,4-Cyclohexanedicarboxylic Acid Dimethyl Ester

Formula: C₁₀H₁₆O₄
MW: 200.2316 g/mol
Boiling Pt: 263 °C
Flash Pt: 94 °C
MDL Number: MFCD00001460
CAS Number: 94-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66faa299118ec0fb3ef07bf105f68dc4e801c440&vid=23580b74dcd1e3274109f8c5ace03285965e6d07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR