2-Cyanopyrazine ≥99.0%

Supplier: TCI America

C1123-25G C1123-500G
TCC1123-025GEA 28.44 CAD
TCC1123-025G TCC1123-500G
2-Cyanopyrazine ≥99.0%
2-Cyanopyrazine

CAS Number: 19847-12-2
MDL Number: MFCD00049361
Molecular Formula: C5H3N3
Molecular Weight: 105.10
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 87
Flash Point (°C): 97
Freezing point (°C): 21

Formula: C₅H₃N₃
Boiling Pt: 87 °C
Flash Pt: 97 °C
MDL Number: MFCD00049361
CAS Number: 19847-12-2
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8351882161de86466c995c834c214eaa77e54a6&vid=e8351882161de86466c995c834c214eaa77e54a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR